CAS RN 583-48-2
IUPAC Name:
3,4-dimethylhexane
Synonyms:
3,4-dimethylhexane
Molecular Weight (g/mol):
114.23
Molecular Formula:
C8H18
InChi Key:
RNTWWGNZUXGTAX-UHFFFAOYNA-N
SMILES:
CCC(C)C(C)CC
1
–
2
of
2
results
1
3,4-Dimethylhexane, 97%
CAS: 583-48-2 Molecular Formula: C8H18 Molecular Weight (g/mol): 114.232 MDL Number: MFCD00039927 InChI Key: RNTWWGNZUXGTAX-UHFFFAOYSA-N Synonym: hexane, 3,4-dimethyl,3,4-dimethyl hexane,acmc-1aquh,c2h5ch ch3 ch ch3 c2h5,3,4-dimethylhexane, analytical standard,3,4-dimethylhexane, mixture of +/-and meso PubChem CID: 11412 IUPAC Name: 3,4-dimethylhexane SMILES: CCC(C)C(C)CC