accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 58249-61-9

NSN

CAS RN 58249-61-9

IUPAC Name: 1,3-benzothiazole-6-carbonitrile
Synonyms: 1,3-benzothiazole-6-carbonitrile
Molecular Weight (g/mol): 160.19
Molecular Formula: C8H4N2S
InChi Key: WYRRTJKOMZONQO-UHFFFAOYSA-N
SMILES: N#CC1=CC=C2N=CSC2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results