accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OOOCH3O

CAS RN 58244-28-3

IUPAC Name: 5-(acetyloxy)-[1,1'-biphenyl]-2-yl acetate
Synonyms: 5-(acetyloxy)-[1,1'-biphenyl]-2-yl acetate
Molecular Weight (g/mol): 270.28
Molecular Formula: C16H14O4
InChi Key: DZVDHXPXHBVBNZ-UHFFFAOYSA-N
SMILES: CC(=O)OC1=CC=C(OC(C)=O)C(=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results