accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 582-83-2

H3COF

CAS RN 582-83-2

IUPAC Name: 1-(4-fluorophenyl)butan-1-one
Synonyms: 1-(4-fluorophenyl)butan-1-one
Molecular Weight (g/mol): 166.2
Molecular Formula: C10H11FO
InChi Key: QHDXPJMOWRLLRV-UHFFFAOYSA-N
SMILES: CCCC(=O)C1=CC=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results