accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COHCH3

CAS RN 58175-57-8

IUPAC Name: 2-propylpentan-1-ol
Synonyms: 2-propylpentan-1-ol
Molecular Weight (g/mol): 130.23
Molecular Formula: C8H18O
InChi Key: LASHFHLFDRTERB-UHFFFAOYSA-N
SMILES: CCCC(CO)CCC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results