accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 581-00-0

IUPAC Name: 2-methyl-4-(morpholin-4-yl)aniline
Synonyms: 2-methyl-4-(morpholin-4-yl)aniline
Molecular Weight (g/mol): 192.26
Molecular Formula: C11H16N2O
InChi Key: ZGJUJDQANIYVAL-UHFFFAOYSA-N
SMILES: CC1=C(N)C=CC(=C1)N1CCOCC1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results