accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 580-16-5

HON

CAS RN 580-16-5

IUPAC Name: quinolin-6-ol
Synonyms: 6-hydroxyquinoline 1H-1,6-epoxyquinoline 6-hdroxyquinoline 6-chinolinol
Molecular Weight (g/mol): 145.16
Molecular Formula: C9H7NO
InChi Key: OVYWMEWYEJLIER-UHFFFAOYSA-N
SMILES: OC1=CC=C2N=CC=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results