accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 5792-36-9

CH3OOHHO

CAS RN 5792-36-9

IUPAC Name: 1-(2,4-dihydroxyphenyl)propan-1-one
Synonyms: 1-(2,4-dihydroxyphenyl)propan-1-one
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: LLBBBYLDTDJMNU-UHFFFAOYSA-N
SMILES: CCC(=O)C1=CC=C(O)C=C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results