accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 578726-67-7

IUPAC Name: 6-(diethylamino)pyridine-3-carbaldehyde
Synonyms: 6-(diethylamino)pyridine-3-carbaldehyde
Molecular Weight (g/mol): 178.24
Molecular Formula: C10H14N2O
InChi Key: PCISUIMHLSRKGR-UHFFFAOYSA-N
SMILES: CCN(CC)C1=NC=C(C=O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results