accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 577-59-3

H3CONOO

CAS RN 577-59-3

IUPAC Name: 1-(2-nitrophenyl)ethan-1-one
Synonyms: 2-nitroacetophenone
Molecular Weight (g/mol): 165.15
Molecular Formula: C8H7NO3
InChi Key: SUGXZLKUDLDTKX-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=CC=C1[N+]([O-])=O
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)