CAS RN 575-38-2
CAS RN 575-38-2
IUPAC Name:
naphthalene-1,7-diol
Synonyms:
napthalene-1,7-diol
Molecular Weight (g/mol):
160.17
Molecular Formula:
C10H8O2
InChi Key:
ZUVBIBLYOCVYJU-UHFFFAOYSA-N
SMILES:
OC1=CC=C2C=CC=C(O)C2=C1
1
–
1
of
1
results
1
1,7-Dihydroxynaphthalene, 97%
CAS: 575-38-2 Molecular Formula: C10H8O2 Molecular Weight (g/mol): 160.172 MDL Number: MFCD00035720 InChI Key: ZUVBIBLYOCVYJU-UHFFFAOYSA-N Synonym: 1,7-dihydroxynaphthalene,1,7-naphthalenediol,napthalene-1,7-diol,unii-7d42f605cs,ccris 7895,1,7 dihydroxynaphthalene,2,8-dihydroxynaphthalene,acmc-1aqz2,unii-m7uz7bfo3a component PubChem CID: 68462 IUPAC Name: naphthalene-1,7-diol SMILES: C1=CC2=C(C=C(C=C2)O)C(=C1)O