accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 57454-67-8

OOOCo

CAS RN 57454-67-8

IUPAC Name: λ²-cobalt(2+) hydrate carbonate
Synonyms: λ²-cobalt(2+) hydrate carbonate
Molecular Weight (g/mol): 136.96
Molecular Formula: CH2CoO4
InChi Key: JLAIPADPFFTYLP-UHFFFAOYSA-L
SMILES: O.[Co++].[O-]C([O-])=O
Quantity 
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)