accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 57381-39-2

FBrN

CAS RN 57381-39-2

IUPAC Name: 2-bromo-5-fluorobenzonitrile
Synonyms: 2-bromo-5-fluorobenzonitrile
Molecular Weight (g/mol): 200.01
Molecular Formula: C7H3BrFN
InChi Key: MDHNVHCZDCSTMS-UHFFFAOYSA-N
SMILES: FC1=CC=C(Br)C(=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results