accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 57219-64-4

ZrOOZrOOOHOOH

CAS RN 57219-64-4

IUPAC Name: zirconium(4+) tetrahydrogen carbonate
Synonyms: zirconium(4+) tetrabicarbonate
Molecular Weight (g/mol): 335.29
Molecular Formula: C4H4O12Zr
InChi Key: BSZKBMAGLBURDO-UHFFFAOYSA-J
SMILES: [Zr+4].OC([O-])=O.OC([O-])=O.OC([O-])=O.OC([O-])=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results