accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 57199-00-5

H3CCH3CH3CH3ClOH3CCH3

CAS RN 57199-00-5

IUPAC Name: 2,4,6-tris(propan-2-yl)benzoyl chloride
Synonyms: 2,4,6-triisopropylbenzoyl chloride
Molecular Weight (g/mol): 266.81
Molecular Formula: C16H23ClO
InChi Key: OSKNTKJPGKHDHV-UHFFFAOYSA-N
SMILES: CC(C)C1=CC(C(C)C)=C(C(Cl)=O)C(=C1)C(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results