accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 57135-09-8

NSSHN

CAS RN 57135-09-8

IUPAC Name: 1H,2H-[1,3]thiazolo[5,4-b]pyridine-2-thione
Synonyms: 1H-[1,3]thiazolo[5,4-b]pyridine-2-thione
Molecular Weight (g/mol): 168.23
Molecular Formula: C6H4N2S2
InChi Key: WITYIAHCBUYCSY-UHFFFAOYSA-N
SMILES: S=C1NC2=CC=CN=C2S1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results