accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COOClCH3

CAS RN 57044-24-3

IUPAC Name: 4-(chloromethyl)-2,2-dimethyl-1,3-dioxolane
Synonyms: 4-(chloromethyl)-2,2-dimethyl-1,3-dioxolane
Molecular Weight (g/mol): 150.6
Molecular Formula: C6H11ClO2
InChi Key: BNPOTXLWPZOESZ-UHFFFAOYNA-N
SMILES: CC1(C)OCC(CCl)O1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results