accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

FBFFFPP(S)(S)(S)(S)CH3CH3H3CH3CCH3CH3CH3CH3Rh

CAS RN 569650-64-2

IUPAC Name: 1-{2-[2,5-bis(propan-2-yl)phospholan-1-yl]phenyl}-2,5-bis(propan-2-yl)phospholane cycloocta-1,5-diene tetrafluoroboranuide rhodium
Synonyms: 1,5-cyclooctadiene 1-[2-(2,5-diisopropylphospholan-1-yl)phenyl]-2,5-diisopropylphospholane rhodium tetrafluoroborate
Molecular Weight (g/mol): 716.48
Molecular Formula: C34H56BF4P2Rh
InChi Key: IJKOQQHIAGRJHA-UHFFFAOYNA-N
SMILES: [Rh].F[B-](F)(F)F.C1CC=CCCC=C1.CC(C)C1CCC(C(C)C)P1C1=CC=CC=C1P1C(CCC1C(C)C)C(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results