accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 5683-41-0

H3CSCH3CH3N

CAS RN 5683-41-0

IUPAC Name: 2,5,6-trimethyl-1,3-benzothiazole
Synonyms: 2,5,6-trimethyl-1,3-benzothiazole
Molecular Weight (g/mol): 177.27
Molecular Formula: C10H11NS
InChi Key: ZZLZXZFVVOXVRG-UHFFFAOYSA-N
SMILES: CC1=NC2=CC(C)=C(C)C=C2S1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results