accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COOOH

CAS RN 56777-24-3

IUPAC Name: benzyl 2-hydroxypropanoate
Synonyms: benzyl 2-hydroxypropanoate
Molecular Weight (g/mol): 180.2
Molecular Formula: C10H12O3
InChi Key: ZYTLPUIDJRKAAM-UHFFFAOYNA-N
SMILES: CC(O)C(=O)OCC1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results