accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 5676-58-4

H3COCH3N

CAS RN 5676-58-4

IUPAC Name: 2,5-dimethyl-1,3-benzoxazole
Synonyms: benzoxazole, 2,5-dimethyl-
Molecular Weight (g/mol): 147.18
Molecular Formula: C9H9NO
InChi Key: XVQGFGKAPKEUFT-UHFFFAOYSA-N
SMILES: CC1=NC2=CC(C)=CC=C2O1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results