accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NH2OHOF

CAS RN 56741-33-4

IUPAC Name: 5-amino-2-fluorobenzoic acid
Synonyms: 5-amino-2-fluorobenzoic acid
Molecular Weight (g/mol): 155.13
Molecular Formula: C7H6FNO2
InChi Key: WYGAIOJWQDRBRJ-UHFFFAOYSA-N
SMILES: NC1=CC(C(O)=O)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results