CAS RN 56578-18-8
CAS RN 56578-18-8
IUPAC Name:
(5E)-dec-5-en-1-ol
Synonyms:
decan-5-en-1-ol
Molecular Weight (g/mol):
156.27
Molecular Formula:
C10H20O
InChi Key:
WYPQHXVMNVEVEB-AATRIKPKSA-N
SMILES:
CCCC\C=C\CCCCO
1
–
1
of
1
results
1
trans-5-Decen-1-ol, 96%
CAS: 56578-18-8 Molecular Formula: C10H20O Molecular Weight (g/mol): 156.27 MDL Number: MFCD00056187 InChI Key: WYPQHXVMNVEVEB-AATRIKPKSA-N Synonym: trans-5-decen-1-ol,e-5-decen-1-ol,trans-5-decenol,5-decen-1-ol, 5e,5-decen-1-ol, e,decan-5-en-1-ol,e-5-decenol,unii-m709v74ozv,5-decen-1-ol,epa pesticide chemical code 078038 PubChem CID: 5283292 IUPAC Name: (E)-dec-5-en-1-ol SMILES: CCCC\C=C\CCCCO