CAS RN 5657-17-0
CAS RN 5657-17-0
IUPAC Name:
disodium 2-({2-[(carboxylatomethyl)amino]ethyl}amino)acetate
Synonyms:
disodium 2-({2-[(carboxylatomethyl)amino]ethyl}amino)acetate
Molecular Weight (g/mol):
220.14
Molecular Formula:
C6H10N2Na2O4
InChi Key:
QMIFISRIAJBCGZ-UHFFFAOYSA-L
SMILES:
[Na+].[Na+].[O-]C(=O)CNCCNCC([O-])=O
1
–
1
of
1
results
1
Thermo Scientific Chemicals Ethylenediamine-N,N'-diacetic acid
CAS: 5657-17-0 Molecular Formula: C6H10N2Na2O4 Molecular Weight (g/mol): 220.14 MDL Number: MFCD00004281 InChI Key: QMIFISRIAJBCGZ-UHFFFAOYSA-L Synonym: edda,ethylenediamine-n,n'-diacetic acid,n,n'-ethylenediglycine,glycine, n,n'-1,2-ethanediylbis,ethylenediaminediacetic acid,glycine, n,n'-ethylenedi,unii-4lx0u5342u,2-2-carboxymethylamino ethylamino acetic acid,ethylenediamine-n,n'-diaceticacid,ethylenediamine-n,n'-di acetic acid PubChem CID: 61975 SMILES: [Na+].[Na+].[O-]C(=O)CNCCNCC([O-])=O