CAS RN 5655-61-8
CAS RN 5655-61-8
IUPAC Name:
(1S,2R,4S)-1,7,7-trimethylbicyclo[2.2.1]heptan-2-yl acetate
Synonyms:
L-bornyl acetate
Molecular Weight (g/mol):
196.29
Molecular Formula:
C12H20O2
InChi Key:
KGEKLUUHTZCSIP-HOSYDEDBSA-N
SMILES:
CC(=O)O[C@@H]1C[C@@H]2CC[C@@]1(C)C2(C)C
1
–
2
of
2
results
1
L-Bornyl acetate, 95%
CAS: 5655-61-8 Molecular Formula: C12H20O2 Molecular Weight (g/mol): 196.29 MDL Number: MFCD00867808,MFCD00135942,MFCD00135942 InChI Key: KGEKLUUHTZCSIP-HOSYDEDBSA-N Synonym: bornyl acetate,-bornyl acetate PubChem CID: 44630108 IUPAC Name: [(1R,3S)-4,7,7-trimethyl-3-bicyclo[2.2.1]heptanyl] acetate SMILES: CC(=O)O[C@@H]1C[C@@H]2CC[C@@]1(C)C2(C)C
2
L-(-)-Bornyl acetate, 95%
CAS: 5655-61-8 Molecular Formula: C12H20O2 Molecular Weight (g/mol): 196.29 MDL Number: MFCD00867808,MFCD00135942,MFCD00135942 InChI Key: KGEKLUUHTZCSIP-HOSYDEDBSA-N Synonym: bornyl acetate,-bornyl acetate PubChem CID: 44630108 SMILES: CC(=O)O[C@@H]1C[C@@H]2CC[C@@]1(C)C2(C)C