accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 563-79-1

CH3CH3H3CH3C

CAS RN 563-79-1

IUPAC Name: 2,3-dimethylbut-2-ene
Synonyms: 2,3-dimethyl-2-butene
Molecular Weight (g/mol): 84.16
Molecular Formula: C6H12
InChi Key: WGLLSSPDPJPLOR-UHFFFAOYSA-N
SMILES: CC(C)=C(C)C
Molecular Weight (g/mol) 
  • (3)
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (4)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (4)