accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 562-90-3

CH3OOSiOCH3OOCH3OOCH3O

CAS RN 562-90-3

IUPAC Name: tris(acetyloxy)silyl acetate
Synonyms: silicon tetraacetate
Molecular Weight (g/mol): 264.26
Molecular Formula: C8H12O8Si
InChi Key: YZVRVDPMGYFCGL-UHFFFAOYSA-N
SMILES: CC(=O)O[Si](OC(C)=O)(OC(C)=O)OC(C)=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results