accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 5604-46-6

NH2NClOClN

CAS RN 5604-46-6

IUPAC Name: 2-amino-4,6-dichloropyrimidine-5-carbaldehyde
Synonyms: 2-amino-4,6-dichloropyrimidine-5-carbaldehyde
Molecular Weight (g/mol): 192
Molecular Formula: C5H3Cl2N3O
InChi Key: GOJUJUVQIVIZAV-UHFFFAOYSA-N
SMILES: NC1=NC(Cl)=C(C=O)C(Cl)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results