accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 55985-85-8

POOHBrBrBr

CAS RN 55985-85-8

IUPAC Name: tris((2-carboxyethyl)triphenylphosphanium) tribromide
Synonyms: tris((2-carboxyethyl)triphenylphosphanium) tribromide
Molecular Weight (g/mol): 1245.8
Molecular Formula: C63H60Br3O6P3
InChi Key: SZKHVYGCMPETHP-UHFFFAOYSA-N
SMILES: [Br-].[Br-].[Br-].OC(=O)CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1.OC(=O)CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1.OC(=O)CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results