accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 5586-88-9

CH3OCl

CAS RN 5586-88-9

IUPAC Name: 1-(4-chlorophenyl)propan-2-one
Synonyms: 1-(4-chlorophenyl)propan-2-one
Molecular Weight (g/mol): 168.62
Molecular Formula: C9H9ClO
InChi Key: WEJRYKSUUFKMBC-UHFFFAOYSA-N
SMILES: CC(=O)CC1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results