accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 55842-56-3

IUPAC Name: methyl 1,3-thiazole-2-carboxylate
Synonyms: methyl 1,3-thiazole-2-carboxylate
Molecular Weight (g/mol): 143.16
Molecular Formula: C5H5NO2S
InChi Key: YRMZUOPMHMPNPG-UHFFFAOYSA-N
SMILES: COC(=O)C1=NC=CS1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results