accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 55750-53-3

ONOOOH

CAS RN 55750-53-3

IUPAC Name: 6-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)hexanoic acid
Synonyms: N-maleoyl-6-aminohexanoic acid
Molecular Weight (g/mol): 211.22
Molecular Formula: C10H13NO4
InChi Key: WOJKKJKETHYEAC-UHFFFAOYSA-N
SMILES: OC(=O)CCCCCN1C(=O)C=CC1=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results