accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OClO

CAS RN 55746-03-7

IUPAC Name: 3,4-dihydro-2H-1-benzopyran-6-carbonyl chloride
Synonyms: 3,4-dihydro-2H-1-benzopyran-6-carbonyl chloride
Molecular Weight (g/mol): 196.63
Molecular Formula: C10H9ClO2
InChi Key: DKSNWGZAMLPCLZ-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CC2=C(OCCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results