accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OO

CAS RN 55745-97-6

IUPAC Name: 3,4-dihydro-2H-1-benzopyran-6-carbaldehyde
Synonyms: 3,4-dihydro-2H-1-benzopyran-6-carbaldehyde
Molecular Weight (g/mol): 162.19
Molecular Formula: C10H10O2
InChi Key: YIHDTNNFJDJYRG-UHFFFAOYSA-N
SMILES: O=CC1=CC2=C(OCCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results