CAS RN 55717-45-8
IUPAC Name:
6-boranylpyridin-3-ol
Synonyms:
6-boranylpyridin-3-ol
Molecular Weight (g/mol):
106.92
Molecular Formula:
C5H6BNO
InChi Key:
ADHMWMAEKRZWPB-UHFFFAOYSA-N
SMILES:
BC1=CC=C(O)C=N1
1
–
1
of
1
results
1
2-Bromo-5-hydroxypyridine, 95%
CAS: 55717-45-8 Molecular Formula: C5H6BNO Molecular Weight (g/mol): 106.92 MDL Number: MFCD04114184 InChI Key: ADHMWMAEKRZWPB-UHFFFAOYSA-N Synonym: 2-bromo-5-hydroxypyridine,6-bromo-3-pyridinol,6-bromo-3-hydroxypyridine,3-pyridinol, 6-bromo,5-hydroxy-2-bromopyridine,pubchem5967,zlchem 1240,2-bromopyridine-5-ol,acmc-209lor,3-hydroxy-6-bromopyridine PubChem CID: 2762865 IUPAC Name: 6-bromopyridin-3-ol SMILES: BC1=CC=C(O)C=N1