accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 55676-22-7

H3CONCl

CAS RN 55676-22-7

IUPAC Name: 1-(6-chloropyridin-3-yl)ethan-1-one
Synonyms: 1-(6-chloropyridin-3-yl)ethanone
Molecular Weight (g/mol): 155.58
Molecular Formula: C7H6ClNO
InChi Key: UXSNZYGTQTXRAD-UHFFFAOYSA-N
SMILES: CC(=O)C1=CN=C(Cl)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results