accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 55628-54-1

O(R)O(S)O(S)O

CAS RN 55628-54-1

IUPAC Name: (2R,3S,4S)-3,4-bis(benzyloxy)-2-[(benzyloxy)methyl]-3,4-dihydro-2H-pyran
Synonyms: (2R,3S,4S)-3,4-bis(benzyloxy)-2-[(benzyloxy)methyl]-3,4-dihydro-2H-pyran
Molecular Weight (g/mol): 416.52
Molecular Formula: C27H28O4
InChi Key: MXYLLYBWXIUMIT-VJGNERBWSA-N
SMILES: C(OCC1=CC=CC=C1)[C@H]1OC=C[C@H](OCC2=CC=CC=C2)[C@@H]1OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results