accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 556-02-5

(R)ONH2OHHO

CAS RN 556-02-5

IUPAC Name: (2R)-2-amino-3-(4-hydroxyphenyl)propanoic acid
Synonyms: (.+-.)-tyrosine para tyrosine L tyrosine p-tyrosine tyrosine, L isomer L-p-tyrosine tyrosine, L- para-tyrosine tyrosinum L-isomer tyrosine
Molecular Weight (g/mol): 181.19
Molecular Formula: C9H11NO3
InChi Key: OUYCCCASQSFEME-MRVPVSSYSA-N
SMILES: N[C@H](CC1=CC=C(O)C=C1)C(O)=O
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)