accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3N

CAS RN 5558-67-8

IUPAC Name: 2,2-diphenylpropanenitrile
Synonyms: 2,2-diphenylpropanenitrile
Molecular Weight (g/mol): 207.28
Molecular Formula: C15H13N
InChi Key: DPVHBXFSKLKYIQ-UHFFFAOYSA-N
SMILES: CC(C#N)(C1=CC=CC=C1)C1=CC=CC=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results