accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

HOBOHO

CAS RN 55552-70-0

IUPAC Name: (furan-3-yl)boronic acid
Synonyms: furan-3-ylboronic acid
Molecular Weight (g/mol): 111.89
Molecular Formula: C4H5BO3
InChi Key: CYEFKCRAAGLNHW-UHFFFAOYSA-N
SMILES: OB(O)C1=COC=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results