accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H2CSOO

CAS RN 5535-48-8

IUPAC Name: (ethenesulfonyl)benzene
Synonyms: (ethenesulfonyl)benzene
Molecular Weight (g/mol): 168.21
Molecular Formula: C8H8O2S
InChi Key: UJTPZISIAWDGFF-UHFFFAOYSA-N
SMILES: C=CS(=O)(=O)C1=CC=CC=C1
Quantity 
  • (3)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results