accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 5524-05-0

CH3(R)(R)OCH3H2C

CAS RN 5524-05-0

IUPAC Name: (2R,5R)-2-methyl-5-(prop-1-en-2-yl)cyclohexan-1-one
Synonyms: (+)-dihydrocarvone (1R,4R)-dihydrocarvone d-dihydrocarvone
Molecular Weight (g/mol): 152.24
Molecular Formula: C10H16O
InChi Key: AZOCECCLWFDTAP-RKDXNWHRSA-N
SMILES: C[C@@H]1CC[C@H](CC1=O)C(C)=C
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results