accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

SSOOClNO

CAS RN 551930-53-1

IUPAC Name: 5-(1,2-oxazol-5-yl)thiophene-2-sulfonyl chloride
Synonyms: 5-(1,2-oxazol-5-yl)thiophene-2-sulfonyl chloride
Molecular Weight (g/mol): 249.68
Molecular Formula: C7H4ClNO3S2
InChi Key: SSGKBJJYLTYNQD-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=CC=C(S1)C1=CC=NO1
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)