accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COOOCH3

CAS RN 54916-28-8

IUPAC Name: 1-[4-(4-methoxyphenoxy)phenyl]ethan-1-one
Synonyms: 1-[4-(4-methoxyphenoxy)phenyl]ethanone
Molecular Weight (g/mol): 242.27
Molecular Formula: C15H14O3
InChi Key: ICXPZMQQZWKKMN-UHFFFAOYSA-N
SMILES: COC1=CC=C(OC2=CC=C(C=C2)C(C)=O)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results