accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 54714-10-2

IUPAC Name: 4-hydroxy-2,5-diphenyl-2,3-dihydro-1λ⁶-thiophene-1,1,3-trione
Synonyms: 4-hydroxy-2,5-diphenyl-2H-1λ⁶-thiophene-1,1,3-trione
Molecular Weight (g/mol): 300.33
Molecular Formula: C16H12O4S
InChi Key: UIVWXNPUCAHAJX-UHFFFAOYNA-N
SMILES: OC1=C(C2=CC=CC=C2)S(=O)(=O)C(C1=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results