accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 547-63-7

CH3OOCH3H3C

CAS RN 547-63-7

IUPAC Name: methyl 2-methylpropanoate
Synonyms: methyl isobutyrate
Molecular Weight (g/mol): 102.13
Molecular Formula: C5H10O2
InChi Key: BHIWKHZACMWKOJ-UHFFFAOYSA-N
SMILES: COC(=O)C(C)C
Boiling Point 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)