accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

ClSNS

CAS RN 54679-16-2

IUPAC Name: 4-(chloromethyl)-2-(thiophen-2-yl)-1,3-thiazole
Synonyms: 4-(chloromethyl)-2-(thiophen-2-yl)-1,3-thiazole
Molecular Weight (g/mol): 215.71
Molecular Formula: C8H6ClNS2
InChi Key: CGJJBHKYGNSTDK-UHFFFAOYSA-N
SMILES: ClCC1=CSC(=N1)C1=CC=CS1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results