CAS RN 545394-83-0
CAS RN 545394-83-0
IUPAC Name:
2-methyl-6-(trifluoromethyl)pyridine-3-carbaldehyde
Synonyms:
2-methyl-6-(trifluoromethyl)pyridine-3-carbaldehyde
Molecular Weight (g/mol):
189.14
Molecular Formula:
C8H6F3NO
InChi Key:
HHGVNSFXEFHMGQ-UHFFFAOYSA-N
SMILES:
CC1=C(C=O)C=CC(=N1)C(F)(F)F
1
–
2
of
2
results
2
2-Methyl-6-(trifluoromethyl)pyridine-3-carboxaldehyde, 97%, Thermo Scientific Chemicals
CAS: 545394-83-0 Molecular Formula: C8H6F3NO Molecular Weight (g/mol): 189.14 MDL Number: MFCD11035943 InChI Key: HHGVNSFXEFHMGQ-UHFFFAOYSA-N Synonym: 2-methyl-6-trifluoromethyl nicotinaldehyde,2-methyl-6-trifluoromethyl pyridine-3-carbaldehyde,2-methyl-6-trifluoromethyl pyridine-3-carboxaldehyde,2-methyl-6-trifluoromethyl-3-pyridinecarboxaldehyde,3-pyridinecarboxaldehyde, 2-methyl-6-trifluoromethyl PubChem CID: 28875322 IUPAC Name: 2-methyl-6-(trifluoromethyl)pyridine-3-carbaldehyde SMILES: CC1=C(C=O)C=CC(=N1)C(F)(F)F