accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 5452-35-7

NH2

CAS RN 5452-35-7

IUPAC Name: cycloheptanamine
Synonyms: cycloheptanamine
Molecular Weight (g/mol): 113.2
Molecular Formula: C7H15N
InChi Key: VXVVUHQULXCUPF-UHFFFAOYSA-N
SMILES: NC1CCCCCC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results