CAS RN 54507-44-7
IUPAC Name:
2-iodo-4-(trifluoromethyl)benzoic acid
Synonyms:
2-iodo-4-(trifluoromethyl)benzoic acid
Molecular Weight (g/mol):
316.02
Molecular Formula:
C8H4F3IO2
InChi Key:
WCLYLTXCKMZNOH-UHFFFAOYSA-N
SMILES:
OC(=O)C1=C(I)C=C(C=C1)C(F)(F)F
1
–
1
of
1
results
1
2-Iodo-4-(trifluoromethyl)benzoic acid, 98%, Thermo Scientific™
CAS: 54507-44-7 Molecular Formula: C8H4F3IO2 Molecular Weight (g/mol): 316.018 MDL Number: MFCD06408817 InChI Key: WCLYLTXCKMZNOH-UHFFFAOYSA-N Synonym: 2-iodo-4-trifluoromethyl benzoic acid,2-iodo-4-trifluoromethyl-benzoic acid,benzoic acid, 2-iodo-4-trifluoromethyl,2-iodo-4-trifluoromethylbenzoicacid,2-iodo-4-trifluoromethyl-benzoicacid PubChem CID: 16244090 IUPAC Name: 2-iodo-4-(trifluoromethyl)benzoic acid SMILES: C1=CC(=C(C=C1C(F)(F)F)I)C(=O)O